Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTRF1L Monoclonal Antibody (OTI2C6), TrueMAB™, OriGene
SDP

Catalog No. 502542562
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2562 30 μL
50-167-2209 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2562 Supplier OriGene Supplier No. TA501066S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MTRF1L Monoclonal antibody specifically detects MTRF1L in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen MTRF1L
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2C6
Concentration 0.59 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene MTRF1L
Gene Accession No. Q9UGC7
Gene Alias 9130004K12Rik; HMRF1L; mitochondrial release factor 1 like; mitochondrial translational release factor 1 like; mitochondrial translational release factor 1-like; MRF1L; mtRF1a; MTRF1L; Peptide chain release factor 1-like, mitochondrial
Host Species Mouse
Immunogen Full length human recombinant protein of human MTRF1L produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54516
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.