Learn More
MTRF1L Mouse anti-Human, Clone: OTI2C6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF501066

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Mitochondrial peptide chain release factor that directs the termination of translation in response to the peptide chain termination codons UAA and UAG.Specifications
MTRF1L | |
Monoclonal | |
Unconjugated | |
MTRF1L | |
HMRF1L, MRF1L, mtRF1a | |
Mouse | |
Affinity Chromatography | |
RUO | |
54516 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI2C6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9UGC7 | |
MTRF1L | |
Full length human recombit protein of human MTRF1L produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.