Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTRR Antibody, Novus Biologicals™
SDP

Catalog No. p-7107696 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP157749 100 μL
NBP15774920 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP157749 Supplier Novus Biologicals Supplier No. NBP157749
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MTRR Polyclonal specifically detects MTRR in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen MTRR
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q9UBK8
Gene Alias [methionine synthase]-cobalamin methyltransferase (cob(II)alamin reducing), 5-methyltetrahydrofolate-homocysteine methyltransferase reductase, cblE, EC 1.16.1.8, methionine synthase reductase, methionine synthase reductase, mitochondrial, MGC129643, MSR
Gene Symbols MTRR
Host Species Rabbit
Immunogen Synthetic peptides corresponding to MTRR (5-methyltetrahydrofolate-homocysteine methyltransferase reductase) The peptide sequence was selected from the N terminal of MTRR)(50ug). Peptide sequence YDLKTETAPLVVVVSTTGTGDPPDTARKFVKEIQNQTLPVDFFAHLRYGL The peptide sequence for this immunogen was taken from within the described region.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4552
Test Specificity Expected identity based on immunogen sequence: Chicken: 100%; Canine: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Bovine: 92%; Guinea pig: 92%; Equine: 92%; Pig: 92%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.