Learn More
Liofilchem™ MTS™ Ertapenem / Ertapenem + Phenylboronic Acid [ETP/EBO] 0.125-8 / 0.032-2 μg/mL
Shop All Liofilchem Products

Description
Specifications
Specifications
| Antimicrobial Agents | Ertapenem / Ertapenem + Phenylboronic Acid |
| Concentration | 0.125 to 8 / 0.032 to 2 μg/mL |
| Content And Storage | -20°C or 2°C to 8°C until the given expiry date |
| Description | Liofilchem™ MTS™ Ertapenem / Ertapenem + Phenylboronic Acid |
| Antibiotic Code | ETP/EBO |
| Product Type | MIC Test Strip |
| For Use With (Application) | Antimicrobial Resistance Detection, For determining the MIC of antimicrobial agents against microorganisms |
| Packaging Type | Individually Packed |
| Quantity | 30/Pk. |
Non-hazardous according to current legislation, but falls within the specific field of application where a safety data sheet must be supplied because it can cause phenomena of sensitization in sensitive subjects if it comes into contact with the skin.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.