Learn More
Invitrogen™ MTSS1 Polyclonal Antibody

Description
The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_b), 94% homologous in monkey, and 70% homologous in rat.
Specifications
Specifications
| Antigen | MTSS1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% sodium azide |
| Gene | MTSS1 |
| Gene Accession No. | O43312, Q8R1S4 |
| Gene Alias | 2310003N14Rik; actin monomer-binding protein; BC024131; D130001D01Rik; hypothetical protein; KIAA0429; metastasis suppressor 1; metastasis suppressor protein 1; Metastasis suppressor YGL-1; Mim; MIMA; MIMB; missing in metastasis protein; mKIAA0429; MTSS I-BAR domain containing 1; MTSS1; MTSS1, I-BAR domain containing; Protein MTSS 1; protein MTSS 1; metastasis suppressor protein 1 |
| Gene Symbols | MTSS1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.