Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTSS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116535
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116535 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116535 Supplier Invitrogen™ Supplier No. PA5116535
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HeLa, HT-29, U-87MG, Mouse liver, Mouse brain, Mouse heart, Rat liver Immunogen sequence: LPAPPDGPEE RGEHSPESPS VGEGPQGVTS MPSSMWSGQA SVNPPLPGPK PSIPEEHRQA IPESEAEDQE REPPSATVSP GQIPESDPAD LSPRDTPQGE D.

Metastasis suppressor protein 1 (MTSS1) [MIM, missing in metastasis], was first discovered as missing transcripts from metastatic bladder and prostate cancer cells. MTSS1 is a 755 amino acid protein, having multidomain and scaffolding function. It has a proline-rich/SH3 binding region and a serine-rich region with EGF or PDGF/Src consensus sites. It contains anactin-binding Wiskott-Aldrich syndrome protein homology-2 motif (WH2) at the C-terminus while the N-terminal IMD domain (IRSp53-MIM domain) interact with membranes and bundle actin filament. The WH2 domain binds to actin monomers and plays an important role in actin cytoskeleton reorganization. Recently, MIM was found to interact with cortactin via SH3 domains. In addition, MTSS1 enhanced cortactin- and Arp2/3-dependent actin polymerization. It is thus involved in cell motility and morphogenesis and also interacts with sonic hedgehog/Gli signaling in epidermal cells. MTSS1 acts as an important regulator of cell growth and development and is known to express in many tissues, including spleen, thymus, prostate, testis, uterus, colon, and peripheral blood.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTSS1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.88 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MTSS1
Gene Accession No. O43312, Q8R1S4
Gene Alias 2310003N14Rik; actin monomer-binding protein; BC024131; D130001D01Rik; hypothetical protein; KIAA0429; metastasis suppressor 1; metastasis suppressor protein 1; Metastasis suppressor YGL-1; Mim; MIMA; MIMB; missing in metastasis protein; mKIAA0429; MTSS I-BAR domain containing 1; MTSS1; MTSS1, I-BAR domain containing; Protein MTSS 1; protein MTSS 1; metastasis suppressor protein 1
Gene Symbols MTSS1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 630-730 of human MTSS1 (NP_0555663).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 211401, 362918, 9788
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.