Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTSS1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561763
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561763 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561763 Supplier Invitrogen™ Supplier No. PA561763
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PTSDWSKVGS HEQPSGATLQ RRKDRVELLR DTEPGPASGG TLGPSGEEAP RPRMSPATIA AKHGEEVSPA ASDLAMVLTR GLSLEHQKSS R Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.

Involved in plasma membrane dynamics. Potentiated PDGF-mediated formation of membrane ruffles and lamellipodia in fibroblasts, acting via RAC1 activation (PubMed:14752106). May function in actin bundling (PubMed:14752106). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTSS1L
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Mtss1l
Gene Accession No. Q765P7
Gene Alias ABBA; ABBA1; ABBA-1; actin-bundling protein with BAIAP2 homology; actin-bundling with BAIAP2 homology protein 1; LOC92154; metastasis suppressor 1 like; metastasis suppressor 1-like; Mtss1l; MTSS1-like protein; MTSS2; Protein MTSS 2
Gene Symbols Mtss1l
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MTSS1L. Recombinant protein control fragment (Product #RP-98559).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92154
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.