Learn More
MTSS1L Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304708AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Involved in plasma membrane dynamics. Potentiated PDGF-mediated formation of membrane ruffles and lamellipodia in fibroblasts, acting via RAC1 activation (PubMed:14752106). May function in actin bundling (PubMed:14752106).Specifications
MTSS1L | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q6P9S0, Q765P7 | |
Mtss1l | |
Between 675 and 725 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
MTSS1L | |
ABBA1, ABBA-1, actin-bundling protein with BAIAP2 homology, actin-bundling with BAIAP2 homology protein 1, LOC92154, metastasis suppressor 1-like | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
244654, 92154 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.