Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ MTT Reagent
SDP

Catalog No. 48902501 Shop All Novus Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-902-501 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 48-902-501 Supplier R&D Systems™ Supplier No. 48902501
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

R&D Systems™ MTT Reagent yields low background absorbance values in the absence of cells and is stable when stored at 4°C.

  • Contains less than 1% (w/v) MTT (3-(4,5-dimethylthiazolyl-2)-2, 5-diphenyl-tetrazolium bromide (CAS # 298-93-1).
  • MTT is toxic and may cause heritable genetic defects. In case of contact, immediately flush eyes or skin with copious amounts of water. If swallowed, wash out mouth with water provided person is conscious
  • For use with  MTT Cell Proliferation Assays

Specifications

Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Detection Method Colorimetric
Product Line R&D Systems
Quantity 25 mL
Sample Volume 25 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.