Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MTTP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576049
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
MTTP encodes the large subunit of the heterodimeric microsomal triglyceride transfer protein. Protein disulfide isomerase (PDI) completes the heterodimeric microsomal triglyceride transfer protein, which has been shown to play a entral role in lipoprotein assembly. Mutations in MTP can cause abetalipoproteinemia.
Specifications
MTTP | |
Polyclonal | |
Unconjugated | |
MTTP | |
1810043K16Rik; ABL; LOW QUALITY PROTEIN: microsomal triglyceride transfer protein large subunit; MGC149819; MGC149820; microsomal triglyceride transfer protein; microsomal triglyceride transfer protein (large polypeptide, 88kDa); microsomal triglyceride transfer protein B; microsomal triglyceride transfer protein large subunit; microsomal triglyceride transfer protein, large subunit; Mtp; MTTP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17777, 310900, 4547 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
O08601, P55157 | |
MTTP | |
Recombinant full length Human MTTP. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction