Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTUS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542568
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542568 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542568 Supplier Invitrogen™ Supplier No. PA542568
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: QLLACGNTKF EALTVVIQHL LSEREEALKQ HKTLSQELVN LRGELVTAST Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

The protein belongs to the FKBP-type peptidyl-prolyl cis/trans isomerase family. It is located in endoplasmic reticulum and acts as molecular chaperones.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTUS1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MTUS1
Gene Accession No. Q9ULD2
Gene Alias AI481402; angiotensin II AT2 receptor-interacting protein 1; angiotensin-II type 2 receptor-interacting protein; AT2 receptor-binding protein; AT2 receptor-interacting protein; AT2 receptor-interacting protein 1; AT2R binding protein; ATBP; ATBP135; ATIP; Atip1; Atip3b; ATIP4; B430010I23Rik; B430305I03Rik; C85752; Cctsg1; Cctsg1-440; Coiled-coiled tumor suppressor gene 1 protein; coiled-coiled tumor supressor gene 1; DKFZp586D1519; DKFZp686F20243; erythroid differentiation-related; FLJ14295; GK1; growth factor inhibitor; ICIS; KIAA1288; MD44; microtubule associated scaffold protein 1; microtubule associated tumor suppressor 1; microtubule-associated tumor suppressor 1; Microtubule-associated tumor suppressor 1 homolog; mitochondrial tumor suppressor 1; Mitochondrial tumor suppressor 1 homolog; mitochondrial tumor suppressor gene 1; mKIAA1288; MP44; MTSG1; Mtus1; transcription factor MTSG1
Gene Symbols MTUS1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human MTUS1 (aa 924-973).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57509
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.