Learn More
MTUS1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | MTUS1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-2516
![]() |
Bethyl Laboratories, Inc
A302411AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan
MTUS1 contains a C-terminal domain and is able to interact with the angiotension II (AT2) receptor and a large coiled-coil region allowing dimerization. One of the isoforms has been shown to be a mitochondrial protein that acts as a tumor suppressor and partcipates in AT2 signaling pathways. Other isoforms may be nuclear or transmembrane proteins but it has not been determined whether they also participate in AT2 signaling pathways.This gene encodes a protein which contains a C-terminal domain able to interact with the angiotension II (AT2) receptor and a large coiled-coil region allowing dimerization. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene. One of the transcript variants has been shown to encode a mitochondrial protein that acts as a tumor suppressor and partcipates in AT2 signaling pathways. Other variants may encode nuclear or transmembrane proteins but it has not been determined whether they also participate in AT2 signaling pathways.Specifications
MTUS1 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Microtubule associated tumor suppressor 1, ATIP, DKFZp586D1519, DKFZp686F20243, FLJ14295, KIAA1288, MP44, MTSG1, ATBP | |
MTUS1 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9ULD2 | |
57509 | |
Between 1220 and 1270 | |
Primary | |
4° C | |
MTUS1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.