Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MTUS1 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS6076R

Description
The protein belongs to the FKBP-type peptidyl-prolyl cis/trans isomerase family. It is located in endoplasmic reticulum and acts as molecular chaperones.Specifications
MTUS1 | |
Polyclonal | |
Unconjugated | |
MTUS1 | |
AI481402; angiotensin II AT2 receptor-interacting protein 1; Angiotensin-II type 2 receptor-interacting protein; AT2 receptor-binding protein; AT2 receptor-interacting protein; AT2 receptor-interacting protein 1; AT2R binding protein; ATBP; ATBP135; Atip; Atip1; Atip3b; ATIP4; B430010I23Rik; B430305I03Rik; BOS_23776; C85752; Cctsg1; Cctsg1-440; Coiled-coiled tumor suppressor gene 1 protein; coiled-coiled tumor supressor gene 1; DKFZp586D1519; DKFZp686F20243; erythroid differentiation-related; FLJ14295; GK1; growth factor inhibitor; ICIS; Kiaa1288; MD44; microtubule associated scaffold protein 1; microtubule associated tumor suppressor 1; Microtubule-associated tumor suppressor 1; Microtubule-associated tumor suppressor 1 homolog; Mitochondrial tumor suppressor 1; Mitochondrial tumor suppressor 1 homolog; mitochondrial tumor suppressor gene 1; mKIAA1288; MP44; MTSG1; MTUS1; transcription factor MTSG1 | |
Rabbit | |
Protein A | |
RUO | |
57509 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q9ULD2 | |
MTUS1 | |
KLH conjugated synthetic peptide derived from human MTUS1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction