Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598434
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598434 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598434 Supplier Invitrogen™ Supplier No. PA598434
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MTX1 Polyclonal Antibody for Western Blot, ELISA

MTX1 belongs to the metaxin family. It is involved in transport of proteins into the mitochondrion and essential for embryonic development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTX1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.42 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MTX1
Gene Accession No. Q13505
Gene Alias Gcap6; LOW QUALITY PROTEIN: metaxin-1; Metaxin 1; metaxin-1; Mitochondrial outer membrane import complex protein 1; Mtx; MTX1; MTXN; Unknown (protein for MGC:134321)
Gene Symbols MTX1
Host Species Rabbit
Immunogen Recombinant Human Metaxin-1 protein (217-466AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4580
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.