Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTX2 Monoclonal Antibody (OTI2E9), OriGene
SDP

Catalog No. 502543854
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3854 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3854 Supplier OriGene Supplier No. CF811991
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MTX2 Monoclonal antibody specifically detects MTX2 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen MTX2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI2E9
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene ZNF8
Gene Accession No. P17098
Gene Alias 1500012G02Rik; metaxin 2; Metaxin2; metaxin-2; Mitochondrial outer membrane import complex protein 2; MNCb-0780; Mtx2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-242 of human ZNF8 (NP_066575) produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7554
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.