Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556777
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556777 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556777 Supplier Invitrogen™ Supplier No. PA556777
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TLDQVLEDVD QCCQALSQRL GTQPYFFNKQ PTELDALVFG HLYTILTTQL TNDELSEKVK NYSNLLAFCR RIEQHYFEDR G Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 94%.

The protein encoded by this gene is highly similar to the metaxin 2 protein from mouse, which has been shown to interact with the mitochondrial membrane protein metaxin 1. Because of this similarity, it is thought that the encoded protein is peripherally associated with the cytosolic face of the outer mitochondrial membrane, and that it is involved in the import of proteins into the mitochondrion. Alternative splicing results in multiple transcript variants. A related pseudogene has been identified on chromosome 7.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTX2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MTX2
Gene Accession No. O75431
Gene Alias 1500012G02Rik; metaxin 2; Metaxin2; metaxin-2; Mitochondrial outer membrane import complex protein 2; MNCb-0780; Mtx2
Gene Symbols MTX2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MTX2. Recombinant protein control fragment (Product #RP-96072).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10651
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.