Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MTX3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566452
Description
Immunogen sequence: FRLSLGGISPA GQETVDANLQ KLTQLVNKES NLIEKMDDNL RQSPQLPPRK LPTLKLTPAE EENNS Highest antigen sequence identity to the following orthologs - mouse 88%, rat 89%.
Could function in transport of proteins into the mitochondrion.
Specifications
MTX3 | |
Polyclonal | |
Unconjugated | |
MTX3 | |
4930470O13Rik; AA409304; AI853833; AU067765; EG624619; Gm1194; Gm6514; metaxin 3; metaxin-3; mtx3; pleckstrin homology domain containing, family M (with RUN domain) member 2; zgc:109718; zMTX3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
345778 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5HYI7 | |
MTX3 | |
Recombinant Human MTX3. Recombinant protein control fragment (Product #RP-107125). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction