Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ MU 1700 trihydrochloride
SDP

Catalog No. 883250 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
88-325-0 50 mg
88-321-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 88-325-0 Supplier Tocris Bioscience™ Supplier No. 8832/50
Add to Cart
Add to Cart

Potent and selective ALK1/2 inhibitor. Brain-penetrant

MU 1700 trihydrochloride is a potent and selective ALK2 and ALK1 inhibitor (IC50 values are 6 nM and 13 nM, respectively in biochemical assays). It is also a potent ALK6 inhibitor (IC50 = 41 nM). It shows selectivity for ALK over >360 other kinases. It shows favorable pharmacokinetic for in vivo use. MU 1700 trihydrochloride is orally bioavailable. It is cell membrane permeable and brain penetrant.

Specifications

Chemical Name or Material 6-(4-Piperazin-1-ylphenyl)-3-quinolin-4-ylfuro[3,2-b]pyridine trihydrochloride
Quantity 50 mg
Target ALK Inhibitors
Molecular Formula C26H22N4O.3HCl
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.