Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mu-Calpain Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA527682
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA527682 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA527682 Supplier Invitrogen™ Supplier No. PA527682
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A431, Jurkat. Predicted reactivity: Mouse (87%), Rat (87%), Xenopus laevis (86%), Pig (96%), Chicken (89%), Sheep (95%), Rhesus Monkey (99%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The calpains including Calpain 1 (CAPN1), calcium-activated neutral proteases, are nonlysosomal, intracellular cysteine proteases. The mammalian calpains include ubiquitous, stomach-specific, and muscle-specific proteins. The ubiquitous enzymes consist of heterodimers with distinct large, catalytic subunits associated with a common small, regulatory subunit. This gene encodes the large subunit of the ubiquitous enzyme, calpain 1. Several transcript variants encoding two different isoforms have been found for this gene. Diseases associated with CAPN1 include Spastic Paraplegia 76, Autosomal Recessive and Spasticity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mu-Calpain
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.55 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene CAPN1
Gene Accession No. P07384
Gene Alias calcium activated neutral protease; calcium-activated neutral protease 1; calcium-activated neutral proteinase 1; calcium-dependent protease; calpain; calpain 1; calpain 1 large subunit; calpain 1, (mu/I) large subunit; calpain 1, large subunit; calpain mu-type; calpain, large polypeptide L1; calpain-1 catalytic subunit; Calpain-1 large subunit; Calpain-I; Calpain-II; CANP; CANP 1; Canp1; CANPL1; Capa1; Capa-1; Capn1; Cell proliferation-inducing gene 30 protein; cell proliferation-inducing protein 30; Cls1; I79_024883; micromolar calcium-activated neutral protease 1; micromolar calcium-activated neutral protease 1 isoform A; micromolar calcium-activated neutral protease 1 isoform B; micromolar calcium-dependent neutral protease large subunit; micromolar-calpain; mucalpain; mu-calpain; mu-calpin; muCANP; muCL; PIG30; SPG76
Gene Symbols CAPN1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Calpain 1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 823
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.