Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
mu Crystallin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$231.00 - $262.00
Specifications
Antigen | mu Crystallin |
---|---|
Dilution | Western Blot 1:500-1:2000, Immunohistochemistry-Paraffin 1:500-1:2000 |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
mu Crystallin Polyclonal antibody specifically detects mu Crystallin in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
mu Crystallin | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Vision | |
PBS (pH 7.0) | |
1428 | |
IgG | |
Antigen and protein A Affinity-purified |
Western Blot 1:500-1:2000, Immunohistochemistry-Paraffin 1:500-1:2000 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
crystallin, mu, DFNA40NADP-regulated thyroid-hormone binding protein, mu-crystallin homolog, NADP-regulated thyroid-hormone-binding protein, THBP | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human mu Crystallin. | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title