Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
mu Opioid R/OPRM1 Antibody

Rabbit Polyclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NB1001620
Description
mu Opioid R/OPRM1 Polyclonal specifically detects mu Opioid R/OPRM1 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
mu Opioid R/OPRM1 | |
Polyclonal | |
Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100-1:2000, Immunohistochemistry-Frozen 1:2500-1:5000 | |
P33535 | |
OPRM1 | |
Synthetic peptide: NHQLENLEAETAPLP, corresponding to C terminal amino acids 384-398 of Rat Mu Opioid Receptor. | |
0.05 mL | |
Cell Cycle and Replication, GPCR, Neuroscience, Neurotransmission, Signal Transduction | |
4988 | |
Human, Mouse, Rat, Primate | |
Antisera |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
Unconjugated | |
Whole antisera with 0.05% Sodium Azide | |
hMOP, KIAA0403, LMOR, MOP, M-OR-1, MOR-1, MOR1MOR, Mu opiate receptor, Mu opioid receptor, mu-type opioid receptor, opioid receptor, mu 1, OPRM | |
Rabbit | |
Unpurified | |
RUO | |
Primary | |
Mu Opioid Receptor | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction