Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
mu Opioid R/OPRM1 Antibody, DyLight 594 , Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11079878DL594
Description
mu Opioid R/OPRM1 Polyclonal antibody specifically detects mu Opioid R/OPRM1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
mu Opioid R/OPRM1 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
DyLight 594 | |
hMOP, KIAA0403, LMOR, MOP, M-OR-1, MOR-1, MOR1MOR, Mu opiate receptor, Mu opioid receptor, mu-type opioid receptor, opioid receptor, mu 1, OPRM | |
Synthetic peptide comprising residues 386-400 of the human, mouse and rat MOR-1 protein. | |
1 mL | |
Cell Cycle and Replication, GPCR, Neuroscience, Neurotransmission, Signal Transduction | |
4988 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction