Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mu Opioid Receptor Rabbit anti-Human, Mouse, Non-human Primate, Rat, Polyclonal, Neuromics™
Rabbit Polyclonal Antibody
Supplier: Neuromics, Inc RA10104150UL
Description
Mu Opioid Receptor Polyclonal antibody specifically detects Mu Opioid Receptor in Human, Mouse, Non-human Primate, Rat samples. It is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western BlotSpecifications
| Mu Opioid Receptor | |
| Polyclonal | |
| Unconjugated | |
| OPRM1 | |
| BOS_9964; hMOP; KIAA0403; LMOR; MOP; MOP receptor; MOP-R; Mor; MOR1; M-OR-1; MOR-1; MOR-1C; MOR-1O; MORA; Mu opiate receptor; Mu opioid receptor; mu opioid receptor hMOR-1a; mu opioid receptor splice variant mMOR-1Z; mu opioid receptor splice variant rMOR-1S; mu opioid receptor splice variant rMOR-1Z; mu-opioid receptor; muOR; MUOR1; Mu-type opioid receptor; Opioid mu receptor; Opioid Receptor; Opioid receptor B; opioid receptor mu 1; opioid receptor, mu 1; Oprm; OPRM1; Oprrm1; OTTHUMP00000017463; Ror-b | |
| NHQLENLEAETAPLP | |
| RUO | |
| 18390, 25601, 4988 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
| Conc. Not Determined | |
| whole serum with 0.05% sodium azide | |
| P33535, P35372, P42866 | |
| Rabbit | |
| 150 μL | |
| Primary | |
| Human, Mouse, Non-human Primate, Rat | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction