Learn More
MUC-2 Monoclonal Antibody (Ccp58), Zeta
Mouse Monoclonal Antibody Cocktail
Supplier: ZETA CORPORATION Z2151ML

Description
A recommended positive control tissue for this product is Colon adenocarcinoma, however positive controls are not limited to this tissue type. The primary antibody is intended for laboratory professional use in the detection of the corresponding protein in formalin-fixed, paraffin-embedded tissue stained in manual qualitative immunohistochemistry (IHC) testing. This antibody is intended to be used after the primary diagnosis of tumor has been made by conventional histopathology using non-immunological histochemical stains.
MUC-2 Monoclonal antibody specifically detects MUC-2 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
MUC-2 | |
Monoclonal | |
200 μg/mL | |
tris with BSA, NP-40 and <0.1% sodium azide | |
Q02817 | |
Mouse | |
Protein A | |
RUO | |
4583 | |
2-8°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
Ccp58 | |
Unconjugated | |
MUC2 | |
2010015E03Rik; colonic mucin; HH-Muc; intestinal mucin-2; MCM; MLP; muc2; MUC-2; mucin 2; mucin 2, intestinal/tracheal; mucin 2, oligomeric mucus/gel-forming; mucin-2; mucin-like protein; secreted gel-forming mucin; SMUC; wnn | |
Synthetic peptide representing a region on the Muc-2 glycoprotein | |
1 mL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.