Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC-4 Rabbit anti-Human, Clone: ZR201, Zeta Corp

Catalog No. 502238152
Encompass_Preferred
Change view
Click to view available options
Quantity:
7 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-223-8152 7 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-223-8152 Supplier ZETA CORPORATION Supplier No. Z2520RP
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is diluted and in a ready-to-use formulation.

A recommended positive control tissue for this product is Colon, however positive controls are not limited to this tissue type.

Specifications

Antigen MUC-4
Applications Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone ZR201
Concentration 2 μg/mL
Conjugate Unconjugated
Formulation tris with BSA, NP-40 and <0.1% sodium azide
Gene MUC4
Gene Accession No. Q99102
Gene Alias 4933405I11Rik; Ascites sialoglycoprotein; Ascites sialoglycoprotein 1; Ascites sialoglycoprotein 2; ASGP; ASGP1; ASGP-1; ASGP-2; HSA276359; MUC4; MUC-4; mucin 4; mucin 4, ASGP; mucin 4, cell surface associated; mucin 4, tracheobronchial; Mucin4; mucin-4; Mucin-4 alpha chain; Mucin-4 beta chain; Pancreatic adenocarcinoma mucin; pre-sialomucin complex; Psmc; sialomucin ascites sialoglycoprotein-1; sialomucin complex; testis mucin; Tracheobronchial mucin
Gene Symbols MUC4
Host Species Rabbit
Immunogen Recombinant fragment (around aa1500-2000) of human MUC4 protein
Purification Method Protein A
Quantity 7 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4585
Target Species Human
Content And Storage 2-8°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.