Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC1 Antibody (139H2) - Azide and BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$678.13

Specifications

Antigen MUC-1
Clone 139H2
Concentration 1.0 mg/mL
Dilution Flow Cytometry : 0.25 - 0.5 ug/million cells in 0.1ml, ELISA, Immunohistochemistry-Paraffin : 0.5 - 1.0 ug/ml, Immunofluorescence : 1 - 2 ug/ml, CyTOF-ready
Applications Flow Cytometry, ELISA, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP247888A
SDP
View Documents
Novus Biologicals
NBP2478880.1MG
0.1 mg
Each for $678.13
Only null left
Add to Cart
 
NBP247888B
SDP
View Documents
Novus Biologicals
NBP2478880.2MG
0.2 mg N/A N/A N/A
Description

Description

MUC1 Monoclonal specifically detects MUC1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence, CyTOF-ready.
Specifications

Specifications

MUC-1
1.0 mg/mL
Flow Cytometry, ELISA, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF
Unconjugated
Mouse
Cancer, Cellular Markers, Extracellular Matrix
PBS with No Preservative
4582
Human milk-fat globule membranes (HMFGM)
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
139H2
Flow Cytometry : 0.25 - 0.5 ug/million cells in 0.1ml, ELISA, Immunohistochemistry-Paraffin : 0.5 - 1.0 ug/ml, Immunofluorescence : 1 - 2 ug/ml, CyTOF-ready
Monoclonal
Purified
RUO
Human, Mouse
Breast carcinoma-associated antigen DF3, Carcinoma-associated mucin, CD227, CD227 antigen, DF3 antigen, EMA, episialin, H23 antigen, H23AG, KL-6, MAM6, MUC-1, MUC1/ZD, mucin 1, cell surface associated, mucin 1, transmembrane, mucin-1, Peanut-reactive urinary mucin, PEMMUC-1/SEC, PEMT, Polymorphic epithelial mucin, PUMMUC-1/X, tumor associated epithelial mucin, Tumor-associated epithelial membrane antigen, Tumor-associated mucin
MUC1
IgG1 κ
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.