Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC1 Antibody (139H2), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$268.00 - $524.00

Specifications

Antigen MUC-1
Clone 139H2
Concentration 0.2mg/mL
Dilution Flow Cytometry 0.25 - 0.5 ug/million cells in 0.1ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 1 - 2 ug/ml
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP24466600
SDP
View Documents
Novus Biologicals
NBP2446660.02MG
0.02 mg
Each for $268.00
Only null left
Add to Cart
 
NBP24466601
SDP
View Documents
Novus Biologicals
NBP2446660.1MG
0.1 mg
Each for $524.00
Only null left
Add to Cart
 
NBP24466602
SDP
View Documents
Novus Biologicals
NBP2446660.2MG
0.2 mg N/A N/A N/A
Description

Description

Ensure accurate, reproducible results in ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence

MUC1 Monoclonal specifically detects MUC1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications

Specifications

MUC-1
0.2mg/mL
Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
Unconjugated
Mouse
Cancer, Cellular Markers, Extracellular Matrix
10mM PBS and 0.05% BSA with 0.05% Sodium Azide
Breast carcinoma-associated antigen DF3, Carcinoma-associated mucin, CD227, CD227 antigen, DF3 antigen, EMA, episialin, H23 antigen, H23AG, KL-6, MAM6, MUC-1, MUC1/ZD, mucin 1, cell surface associated, mucin 1, transmembrane, mucin-1, Peanut-reactive urinary mucin, PEMMUC-1/SEC, PEMT, Polymorphic epithelial mucin, PUMMUC-1/X, tumor associated epithelial mucin, Tumor-associated epithelial membrane antigen, Tumor-associated mucin
MUC1
IgG1 κ
Protein A or G purified
139H2
Flow Cytometry 0.25 - 0.5 ug/million cells in 0.1ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 1 - 2 ug/ml
Monoclonal
Purified
RUO
Human, Mouse
P15941
4582
Human milk-fat globule membranes (HMFGM)
Primary
Store at 4C.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.