Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MUC1 Antibody (604804), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB6298W
Description
MUC1 Monoclonal antibody specifically detects MUC1 in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOFSpecifications
MUC1 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF | |
604804 | |
Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready | |
Breast carcinoma-associated antigen DF3, Carcinoma-associated mucin, CD227, CD227 antigen, DF3 antigen, EMA, episialin, H23 antigen, H23AG, KL-6, MAM6, MUC-1, MUC1/ZD, mucin 1, cell surface associated, mucin 1, transmembrane, mucin-1, Peanut-reactive urinary mucin, PEMMUC-1/SEC, PEMT, Polymorphic epithelial mucin, PUMMUC-1/X, tumor associated epithelial mucin, Tumor-associated epithelial membrane antigen, Tumor-associated mucin | |
E. coli-derived recombinant human MUC-1, Pro126-Arg145, Accession # P15941 | |
0.1 mL | |
Cancer, Cellular Markers, Extracellular Matrix, Inflammation, Signal Transduction | |
4582 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction