Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MUC1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 4 publications
Supplier: Novus Biologicals NBP160046SS
Description
MUC1 Polyclonal specifically detects MUC1 in Human, Mouse, Rat, Porcine, Bovine, Equine, Guinea Pig, Goat, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
MUC1 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
Breast carcinoma-associated antigen DF3, Carcinoma-associated mucin, CD227, CD227 antigen, DF3 antigen, EMA, episialin, H23 antigen, H23AG, KL-6, MAM6, MUC-1, MUC1/ZD, mucin 1, cell surface associated, mucin 1, transmembrane, mucin-1, Peanut-reactive urinary mucin, PEMMUC-1/SEC, PEMT, Polymorphic epithelial mucin, PUMMUC-1/X, tumor associated epithelial mucin, Tumor-associated epithelial membrane antigen, Tumor-associated mucin | |
Rabbit | |
122 kDa | |
0.025 mg | |
Cancer, Cellular Markers, Extracellular Matrix, Inflammation, Signal Transduction | |
The immunogen for this antibody has 100% homology to human MUC1 isoforms S2, E2, 9, F, T10, 5, Y, Y-LSP, 8, 4, 3, 2, 1 and J13. | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1.0 mg/mL | |
Western Blot 0.2-1 ug/ml, Flow Cytometry reported in scientific literature (PMID 28740504), Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 4-8 ug/ml, Knockout Validated | |
Q7Z552 | |
MUC1 | |
Synthetic peptides corresponding to a C terminal portion of the human MUC1 (between amino acids 1200-1250) [UniProt P15941]. | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Pig, Bovine, Equine, Guinea Pig, Goat, Rabbit | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction