Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MUC1 Antibody (OTI2E3), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272826IR
Description
MUC1 Monoclonal specifically detects MUC1 in Human, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
MUC1 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MUC1 | |
Full length recombinant protein of human MUC-1 (NP_001018016) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Canine, Monkey | |
IgG2a |
Western Blot, Immunohistochemistry | |
OTI2E3 | |
Western Blot, Immunohistochemistry | |
Breast carcinoma-associated antigen DF3, Carcinoma-associated mucin, CD227, CD227 antigen, DF3 antigen, EMA, episialin, H23 antigen, H23AG, KL-6, MAM6, MUC-1, MUC1/ZD, mucin 1, cell surface associated, mucin 1, transmembrane, mucin-1, Peanut-reactive urinary mucin, PEMMUC-1/SEC, PEMT, Polymorphic epithelial mucin, PUMMUC-1/X, tumor associated epithelial mucin, Tumor-associated epithelial membrane antigen, Tumor-associated mucin | |
Mouse | |
Affinity Purified | |
Cancer, Cellular Markers, Extracellular Matrix, Inflammation, Signal Transduction | |
4582.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction