Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC1 (CD227) Mouse anti-Human, Brilliant Violet 605, Clone: HMPV, BD Optibuild™

Catalog No. BDB743407 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB743407 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB743407 Supplier BD Biosciences Supplier No. 743407
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The HMPV monoclonal antibody specifically binds to CD227 which is also known as Mucin-1 (MUC1). A major form of CD227 is expressed as a type I transmembrane glycoprotein. CD227 belongs to the epithelial mucin family whose members are heavily O-glycosylated and characterized by high molecular weight, and an amino acid composition rich in serine, threonine, proline, and glycine. CD227 is variably expressed on the surfaces of normal and malignant glandular and ductal epithelial cells, and some hematopoietic cell lineages including subsets of T cells, B cells, monocytes and dendritic cells. Soluble forms of CD227 may arise by shedding from the cell surface or by secretion of forms derived from alternative RNA splicing. The HMPV antibody binds to the core peptide of the MUC1 protein. The core protein contains a domain of 20 amino-acid tandem repeats which function as multiple epitopes for this monoclonal antibody. Incomplete glycosylation of some tumor-associated mucins may lead to variable unmasking of the multiple peptide epitopes leading to the observed differences in immunostaining intensities between cells from normal and malignant tissues. CD227 plays roles in the provision of protective barrier function, the regulation of cellular adhesion, and the transduction of multiple signal pathways.

This antibody is conjugated to BD Horizon™ BV605 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max of 602nm, BD Horizon BV605 can be excited by a violet laser and detected with a standard 610/20nm filter set. BD Horizon BV605 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an Em max at 605nm. Due to the excitation of the acceptor dye by the green (532 nm) and yellow-green (561 nm) lasers, there will be significant spillover into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers. BD Horizon BV605 conjugates are very bright, often exhibiting brightness equivalent to PE conjugates and can be used as a third color off of the violet laser. BV605 is a tandem fluorochrome that combines BD Horizon BV421 and Cy™3.5. These conjugates are very bright, exhibiting similar brightness to equivalent PE conjugates. Due to the excitation of Cy3.5 by the green (532nm) and yellow-green (561nm) lasers, there will be significant spillover of the BD Horizon BV605 signal into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MUC1 (CD227)
Classification Monoclonal
Clone HMPV
Concentration 0.2mg/mL
Conjugate Brilliant Violet 605
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias MUC1, CA 15-3, DF3, EMA, Episialin, H23, H23AG, KL-6, MAM6, PEM, PEMT, PUM
Host Species Mouse
Immunogen Human Milk Fat Globule Membranes
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.