Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MUC1 (CD227) Mouse anti-Human, BUV395, Clone: HMFG2 (also known as HMFG-2, 3.14.A3 or F314AE11), BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 747650
Description
MUC1 (CD227) Monoclonal antibody specifically detects MUC1 (CD227) in Human samples. It is validated for Flow Cytometry.
Specifications
MUC1 (CD227) | |
Monoclonal | |
0.2 mg/mL | |
Mouse | |
RUO | |
Human | |
Purified |
Flow Cytometry | |
HMFG2 (also known as HMFG-2, 3.14.A3 or F314AE11) | |
Brilliant Ultraviolet 395 | |
50 μg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG1 λ |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction