Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC1 Mouse anti-Human, Clone: HMFG2 , MilliporeSigma™

Catalog No. MABC1613MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC1613MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC1613MI Supplier MilliporeSigma Supplier No. MABC1613
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Mucin-1 using this mouse monoclonal Anti-MUC1, clone HMFG2 Immunohistochemistry (Paraffin), Immunoprecipitation, Functional studies, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MUC1
Applications Immunoprecipitation, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HMFG2
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols MUC1,PUM
Host Species Mouse
Immunogen Delipidated milk fat globule extract followed 3 weeks later by epithelial cells cultured from milk (for 2 weeks).
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis and Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_002447.4
Test Specificity Clone HMFG2 immunostains human breast cancer tissue. It can react with the unglycosylated peptide as well as several glycoforms of MUC1.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.