Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MUC1 Antibody (139H2), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP247888APCCY7
Description
MUC1 Monoclonal specifically detects MUC1 in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
| MUC1 | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| MUC1 | |
| Human milk-fat globule membranes (HMFGM) (Uniprot: P15941 ) | |
| 0.1 mL | |
| Primary | |
| This monoclonal antibody reacts with MUC1. The dominant epitope of this monoclonal antibody has not yet been determined. MUC1 is a large cell surface mucin glycoprotein expressed by most glandular and ductal epithelial cells and some hematopoietic cell lineages. It is expressed on most secretory epithelium, including mammary gland and some hematopoietic cells. It is expressed abundantly in lactating mammary glands and over expressed abundantly in 90% breast carcinomas and metastases. Transgenic MUC1 has been shown to associate with all four c-erbB receptors and localize with c-erbB1 (EGFR) in lactating glands. The MUC1 gene contains seven exons and produces several different alternatively spliced variants. The major expressed form of MUC1 uses all seven exons and is a type 1 transmembrane protein with a large extracellular tandem repeat domain. The tandem repeat domain is highly O glycosylated and alterations in glycosylation have been shown in epithelial cancer cells. Antibody to EMA is useful as a pan-epithelial marker for detecting early metastatic loci of carcinoma in bone marrow or liver. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| 139H2 | |
| Flow Cytometry | |
| Breast carcinoma-associated antigen DF3, Carcinoma-associated mucin, CD227, CD227 antigen, DF3 antigen, EMA, episialin, H23 antigen, H23AG, KL-6, MAM6, MUC-1, MUC1/ZD, mucin 1, cell surface associated, mucin 1, transmembrane, mucin-1, Peanut-reactive urinary mucin, PEMMUC-1/SEC, PEMT, Polymorphic epithelial mucin, PUMMUC-1/X, tumor associated epithelial mucin, Tumor-associated epithelial membrane antigen, Tumor-associated mucin | |
| Mouse | |
| Protein A or G purified | |
| Cancer, Cellular Markers, Extracellular Matrix, Inflammation, Signal Transduction | |
| 4582.0 | |
| Human, Mouse | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction