Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MUC12 Monoclonal Antibody (4A3D3)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA548680
Description
MUC12 Monoclonal Antibody for Western Blot, IHC (P), Flow, ELISA
Epitheial mucins are secreted glycoproteins that play roles in cell protection, adhesion modulation and signaling. MUC12 is a novel mucin gene and may be involved in epithelial cell growth regulation. MUC12 is down-regulated in colorectal cancers.
Specifications
MUC12 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
Q9UKN1 | |
MUC12 | |
Purified recombinant fragment of human MUC12 (AA: extra 371-592) expressed in HEK293-6e cells supernatant. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
4A3D3 | |
Unconjugated | |
MUC12 | |
MUC11; MUC-11; MUC12; MUC-12; mucin 11; mucin 12, cell surface associated; mucin-11; Mucin12; Mucin-12 | |
Mouse | |
Protein G | |
RUO | |
10071 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction