Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MUC13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583762
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583762 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583762 Supplier Invitrogen™ Supplier No. PA583762
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ESTTNVNSLA TSDIITASSP NDGLITMVPS ETQSNNEMSP TTEDNQSSGP PTGTALLETS TLNSTGPSNP CQDDPCADNS LCVKLHNTSF CLCLEGYYYN SST Highest antigen sequence indentity to the following orthologs: Rat - 34%, Mouse - 41%.

Mucin 13, epithelial transmembrane. SEA is an extracellular domain associated with O-glycosylation. Proteins found to contain SEA-modules include, agrin, enterokinase, 63 kDa sea urchin (Strongylocentrotus purpuratus) sperm protein, perlecan (heparan sulphate proteoglycan core, mucin 1 and the cell surface antigen, 114/A10, and two functionally uncharacterised, probably extracellular, Caenorhabditis elegans proteins. Despite the functional diversity of these adhesive proteins, a common denominator seems to be their existence in heavily glycosylated environments. In addition, the better characterised proteins all contain O-glycosidic-linked carbohydrates such as heparan sulphate that contribute considerably to their molecular masses. The common module might regulate or assist binding to neighbouring carbohydrate moieties. Enterokinase, the initiator of intestinal digestion, is a mosaic protease composed of a distinctive assortment of domains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MUC13
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MUC13
Gene Accession No. Q9H3R2
Gene Alias 114/A10; 14/A10; AI159736; Cell surface antigen 114/A10; down-regulated in colon cancer 1; DRCC1; Lrrp; Ly64; Lymphocyte antigen 64; Muc-123; MUC13; MUC-13; mucin 13; mucin 13, cell surface associated; mucin 13, epithelial transmembrane; mucin-13; NJ-1; RECC; UNQ6194/PRO20221
Gene Symbols MUC13
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MUC13. Recombinant protein control fragment (Product #RP-88570).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56667
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.