Learn More
MUC16 Mouse anti-Human, Clone: OTI2F1C9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF801881

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
CA 125 determit is present on a high molecular weight, mucin like glycoprotein of high molecular weight. CA 125 has been found on frozen sections of amnion and derivatives of coelomic and mullerian epithelium, including pleura, pericardium and peritoneum. In adult tissues, epithelial cells of fallopian tube, endometrium and endocervix; pancreas, colon, gall bladder, stomach, kidney, apocrine sweat gland and mammary gland. It is also found in mesothelial cell lining of pleura, pericardium and peritoneum. It is found in ovarian tumors of serous, endometrioid or clear cell types and adenocarcinomas of mullerian type.Specifications
MUC16 | |
Monoclonal | |
Unconjugated | |
MUC16 | |
CA125 | |
Mouse | |
Affinity Chromatography | |
RUO | |
94025 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2F1C9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q8WXI7 | |
MUC16 | |
Human recombit protein fragment corresponding to amino acids 13942-14063 and 21117-21238 of human MUC16 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.