Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MUC2 Antibody (CL9148), Novus Biologicals™

Mouse Monoclonal Antibody
$330.00 - $547.00
Specifications
Antigen | MUC2 |
---|---|
Clone | CL9148 |
Dilution | Immunohistochemistry 1:20000 - 1:50000, Immunohistochemistry-Paraffin 1:20000 - 1:50000 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
MUC2 Monoclonal antibody specifically detects MUC2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
MUC2 | |
Immunohistochemistry 1:20000 - 1:50000, Immunohistochemistry-Paraffin 1:20000 - 1:50000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Intestinal mucin-2, MLP, MUC2, MUC-2, mucin 2, intestinal/tracheal, mucin 2, oligomeric mucus/gel-forming, mucin-2, SMUC | |
This antibody was developed against a Synthetic Peptide of Q02817. Actual immunogen sequence is proprietary. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
CL9148 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cancer, Cellular Markers, Inflammation, Signal Transduction | |
PBS, pH 7.2, 40% glycerol | |
4583 | |
IgG2a | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title