Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MUC2 Antibody (MLP/842), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP247685MFV610
Description
MUC2 Monoclonal antibody specifically detects MUC2 in Human, Rat (Negative) samples. It is validated for Flow CytometrySpecifications
MUC2 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Rat (Negative) | |
Purified |
Flow Cytometry | |
MLP/842 | |
Flow Cytometry | |
Intestinal mucin-2, MLP, MUC2, MUC-2, mucin 2, intestinal/tracheal, mucin 2, oligomeric mucus/gel-forming, mucin-2, SMUC | |
Recombinant fragment covering tandem repeats of human MUC2 protein (exact sequence is proprietary) (Uniprot: Q02817 ) | |
0.1 mL | |
Cancer, Cellular Markers, Inflammation, Signal Transduction | |
4583 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction