Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MUC2 Monoclonal Antibody (C1), Invitrogen™

Catalog No. PIMA543654
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543654 20 μL
PIMA543653 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543654 Supplier Invitrogen™ Supplier No. MA543654
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Secreted epithelial mucins are large macromolecules which exhibit extreme polydispersity. Mucin 2 is the major intestinal mucin. O-glycans are attached to MUC2 in a potentially diverse arrangement, which is crucial for their interaction with endogeneous and exogeneous lectins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MUC2
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone C1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene MUC2
Gene Accession No. Q02817
Gene Alias 2010015E03Rik; colonic mucin; HH-Muc; intestinal mucin-2; MCM; MLP; muc2; MUC-2; mucin 2; mucin 2, intestinal/tracheal; mucin 2, oligomeric mucus/gel-forming; mucin-2; mucin-like protein; secreted gel-forming mucin; SMUC; wnn
Gene Symbols MUC2
Host Species Mouse
Immunogen Recombinant protein Mucin 2. The antigen corresponds to amino acid range Cys5075-Gly5179 of the target protein.
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4583
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.