Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC2 Mouse anti-Human, KHL, Clone: CCP58, MilliporeSigma™

Catalog No. MABF19892MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF19892MI 25 μg
MABF19891MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF19892MI Supplier MilliporeSigma Supplier No. MABF198925UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MUC2 Monoclonal antibody specifically detects MUC2 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MUC2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone CCP58
Conjugate KLH
Gene Alias MUC-2;Intestinal Mucin-2
Gene Symbols MUC2, SMUC
Host Species Mouse
Immunogen KLH-conjugated linear peptide (MI-29) corresponding to 29 amino acids from human Mucin-2 containing one repeat unit of 23 amino acids and part of the next repeat of four amino acids.
Molecular Weight of Antigen 540.30 kDa calculated.
Purification Method Protein G Purified
Quantity 25 μg
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone CCP58 is a mouse monoclonal antibody that specifically detects Mucin-2 derived peptides, but does not react with Mucin-1 and 3 derived peptides.
Target Species Human
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.