Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MUC2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA579702
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - IHC: human colon cancer tissue, mouse colon tissue, rat colon tissue.
Secreted epithelial mucins are large macromolecules which exhibit extreme polydispersity. Mucin 2 is the major intestinal mucin. O-glycans are attached to MUC2 in a potentially diverse arrangement, which is crucial for their interaction with endogeneous and exogeneous lectins.
Specifications
MUC2 | |
Polyclonal | |
Unconjugated | |
MUC2 | |
2010015E03Rik; colonic mucin; HH-Muc; intestinal mucin-2; MCM; MLP; muc2; MUC-2; mucin 2; mucin 2, intestinal/tracheal; mucin 2, oligomeric mucus/gel-forming; mucin-2; mucin-like protein; secreted gel-forming mucin; SMUC; wnn | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17831, 24572, 4583 | |
-20°C | |
Lyophilized |
Immunohistochemistry (Paraffin) | |
500 μg/mL | |
PBS with 4mg trehalose and no preservative | |
Q02817, Q62635, Q80Z19 | |
MUC2 | |
A synthetic peptide corresponding to a sequence of human MUC2 (DDFKTASGLVEATGAGFANTWKAQSTCHDKLDWLDD). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction