Learn More
Invitrogen™ MUC20 Polyclonal Antibody

Description
The antibody detects endogenous levels of total MUC20 protein.
Specifications
Specifications
| Antigen | MUC20 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | Muc20 |
| Gene Accession No. | Q8BUE7, Q8N307 |
| Gene Alias | BC026367; KIAA1359; Muc20; MUC-20; mucin 20; mucin 20, cell surface associated; mucin-20; transmembrane mucin MUC20S; UNQ2782/PRO7170 |
| Gene Symbols | Muc20 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.