Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MUC3 Antibody (MUC3/1154), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP247687MFV500
Description
MUC3 Monoclonal antibody specifically detects MUC3 in Human, Rat (Negative) samples. It is validated for Flow CytometrySpecifications
MUC3 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
4584 | |
Store at 4C in the dark. | |
IgG2a κ |
Flow Cytometry | |
MUC3/1154 | |
Flow Cytometry | |
MUC-3A, mucin 3, intestinal, mucin 3A, cell surface associated, mucin 3A, intestinal, mucin-3A | |
A synthetic peptide of 35 amino acids, SIB35 (C-HSTPSFTSSITTTETTSHSTPSFTSSITTTETTS), which contains two of the MUC3 tandem repeats, coupled to KLH. | |
0.1 mL | |
Primary | |
Human, Rat (Negative) | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction