Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MUC3 Antibody (rMUC3/1154), DyLight 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314126DL594
This item is not returnable.
View return policy
Description
MUC3 Monoclonal antibody specifically detects MUC3 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
MUC3 | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
4584 | |
Store at 4°C in the dark. | |
Purified |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
rMUC3/1154 | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
MUC-3A, mucin 3, intestinal, mucin 3A, cell surface associated, mucin 3A, intestinal, mucin-3A | |
Recombinant fragment of human MUC3A protein (exact sequence is proprietary) | |
0.1 mL | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction