Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC3 Monoclonal Antibody (1143/B7), Invitrogen™

Catalog No. PIMA512350
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA512350 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA512350 Supplier Thermo Scientific Supplier No. MA512350

Mouse Monoclonal Antibody

MA5-12350 targets Mucin 3 in IHC (P) applications and shows reactivity with Human samples. The MA5-12350 immunogen is mUC3 tandem repeat peptide.

MUC3 Monoclonal antibody specifically detects MUC3 in Human samples. It is validated for Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MUC3
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1143/B7
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene MUC3A
Gene Accession No. Q02505
Gene Alias A630081J09Rik; intestinal mucin; Intestinal mucin-3A; MUC3; MUC3A; MUC-3A; mucin 3; mucin 3, intestinal; mucin 3A; mucin 3A, cell surface associated; Mucin-3; mucin-3A; RIKEN cDNA A630081J09 gene
Gene Symbols MUC3A
Host Species Mouse
Immunogen MUC3 tandem repeat peptide
Purification Method Protein A
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4584
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.