Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MUC3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA582409
Description
Immunogen sequence: PCPGTITITI VPASPTDPCV EMDPSTEATS PPTTPLTVFP FTTEMVTCPT SISIQTTLTT YMDTSSMMPE SESSISPNAS SSTGTGTVPT NTVFTSTRLP TSETWLSNSS VIPLPLPGVS TIPLTMKPSS S.
MUC3 is an intestinal glycoprotein distributed in the colon and rectum, and to a lesser extent in breast, lung and salivary tissues. It is involved in conferring mucous visoelasticity (RGD).
Specifications
MUC3 | |
Polyclonal | |
Unconjugated | |
MUC3A | |
A630081J09Rik; intestinal mucin; Intestinal mucin-3A; MUC3; MUC3A; MUC-3A; mucin 3; mucin 3, intestinal; mucin 3A; mucin 3A, cell surface associated; Mucin-3; mucin-3A; RIKEN cDNA A630081J09 gene | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4584 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q02505 | |
MUC3A | |
Recombinant protein corresponding to Human MUC3A. Recombinant protein control fragment (Product #RP-88568). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction