Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC3A Rabbit anti-Human, Unconjugated, Clone: MUC3/2992R, Abnova™

Catalog No. 89339565 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-565 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-565 Supplier Abnova Corporation Supplier No. RAB00586
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

MUC3A Monoclonal antibody specifically detects MUC3A in Human samples. It is validated for IHC-P.

Specifications

Antigen MUC3A
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MUC3/2992R
Conjugate Unconjugated
Description mucin 3A, cell surface associated
Gene MUC3A
Gene Alias MUC3, mucin
Host Species Rabbit
Immunogen Recombinant tag fusion protein corresponding to a 35 amino acid fragment of human MUC3A
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 4584, 57876
Target Species Human
Content And Storage Store at -20 to -80°C
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.