Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MUC3B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
$237.00 - $511.50
Specifications
Antigen | MUC3B |
---|---|
Concentration | 0.5 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5121757
|
Invitrogen™
PA5121757 |
20 μL |
Each for $237.00
|
|
|||||
PIPA5121756
|
Invitrogen™
PA5121756 |
100 μL |
Each for $511.50
|
|
|||||
Description
Major glycoprotein component of a variety of mucus gels. Thought to provide a protective, lubricating barrier against particles and infectious agents at mucosal surfaces.Specifications
MUC3B | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9H195 | |
57876 | |
Recombinant Mucin 3B, Cell Surface Associated (MUC3B), Thr13076-Leu13171 (Accession Q9H195). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
MUC3B |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
MUC3; MUC3A; MUC3B; mucin 3; mucin 3B, cell surface associated; Mucin-3B | |
MUC3B | |
IgG | |
Antigen affinity chromatography, Protein A | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title