Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MUC4 Antibody (781631) [DyLight 680], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB8195Y
Description
MUC4 Monoclonal antibody specifically detects MUC4 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
MUC4 | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
781631 | |
Flow Cytometry, CyTOF-ready | |
Ascites sialoglycoprotein, ASGP, HSA276359, MUC-4, mucin 4, cell surface associated, mucin 4, tracheobronchial, mucin-4, Pancreatic adenocarcinoma mucin, Testis mucin, Tracheobronchial mucin | |
E. coli -derived recombinant human MUC4, Pro1072-Ser1317, Accession # Q99102 | |
0.1 mL | |
Cancer | |
4585 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction